Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Peroxide, 95%

Catalog No. AC350410025
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC350410025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC350410025 Supplier Thermo Scientific Chemicals Supplier No. 350410025
Only null left
Add to Cart

CAS: 1304-29-6 | BaO2 | 169.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1304-29-6
Molecular Formula BaO2
Molecular Weight (g/mol) 169.33
MDL Number MFCD00003454
InChI Key ZJRXSAYFZMGQFP-UHFFFAOYSA-N
Synonym barium peroxide, barium dioxide, barium binoxide, barium superoxide, barium oxide, per, bario perossido di, bariumperoxid german, bariumperoxyde dutch, dioxyde de baryum french, barium peroxide ba o2
PubChem CID 14773
IUPAC Name barium(2+);peroxide
SMILES [O-][O-].[Ba+2]

Specifications

Melting Point 450.0°C
CAS Min % 94.0
CAS Max % 100.0
Quantity 2.5 kg
Assay Percent Range 94% min. (Iodometry)
Linear Formula BaO2
Merck Index 15,985
Solubility Information Solubility in water: 0.5g/L
Formula Weight 169.33
Percent Purity 95%
Packaging Plastic bottle
Chemical Name or Material Barium Peroxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
EINECSNumber 215-128-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.