Learn More
Barium peroxide, anhydrous, 84% min
Description
Barium peroxide is a specialty product for proteomics research. It is a strong oxidizing agent. It is also used for bleaching in a variety of commercial applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 450°C (decomposition) |
| Color | White |
| Physical Form | Powder |
| Quantity | 5 kg |
| Assay Percent Range | 84% |
| Solubility Information | Insoluble in water |
| Formula Weight | 169.34 |
| Odor | Odorless |
| Sensitivity | Moisture sensitive |
| Density | 4.96 |
| Show More |
Safety and Handling
| Hazard Category | H272-H302+H332 |
| Hazard Statement | Keep away from food, drink and animal feeding stuffs. Avoid contact with skin and eyes. Take off immediately all contaminated clothing. |
| Precautionary Statement | P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P501c |
| DOTInformation | DOT Class: 5.1, Packaging Group: II |
| EINECSNumber | 215-128-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.