Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Barium sulfate, Puratronic™, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 1065122

Quantity 100g

 View more versions of this product

Catalog No. AA1065122


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7727-43-7
233.4
14,994
Puratronic™
BaSO4
MFCD00003455
Barium sulfate

Specifications

7727-43-7
BaSO4
MFCD00003455
4.5g/mL
White
100g
99.998%, (metals basis)
Powder
233.4
14,994
Odorless
1580°C (dec.)
Barium sulfate
Puratronic™

Safety and Handling

SDS & Safety Documents

RTECSNumber : CR0600000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit