Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium sulfate, Puratronic™, 99.998% (metals basis)

Catalog No. p-7048475
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 7727-43-7 | BaO4S | 233.38 g/mol

Barium sulfate has wide range of applications. It is primarily used in oil well drilling fluid, and as filler in the paint and varnish industry. It is one of the major constituents of the pigment Lithiopone and the white pigment for paints. It improves the resistance against acid and acidity for plastics when employed as filler. Further it is employed as a component in strobe pyrotechnic compositions, & as a coating for photographic papers to improve the effectiveness of the image. Baryta, a thin layer of barium sulfate, is used to make the papers bright for ink-jet printing. In the chemical and pharmaceutical industry, it is used as a catalyst support when selective hydrogenations are required, for e.g. in Rosenmund reduction (Pd on barium sulfate). In colorimetry, barium sulfate is used as a diffuser to measure light sources. In the medical field, barium sulfate suspension is used clinically as a radiocontrast agent for X-ray imaging and other diagnostic procedures. In biomaterials research, it is one of the components utilized in the preparation of acrylic bone cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-43-7
Molecular Formula BaO4S
Molecular Weight (g/mol) 233.38
MDL Number MFCD00003455
InChI Key TZCXTZWJZNENPQ-UHFFFAOYSA-L
Synonym barium sulfate, barite, baritop, barosperse, barotrast, esophotrast, liquipake, macropaque, micropaque, microtrast
PubChem CID 24414
SMILES [Ba++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1580°C (decomposition)
Color White
Physical Form Powder
Quantity 5 g
Linear Formula BaSO4
Merck Index 14,994
Solubility Information Insoluble in water; soluble in hot concentrated sulphuric acid.
IUPAC Name barium(2+);sulfate
Formula Weight 233.4
Percent Purity 99.998%
Grade Puratronic™
Odor Odorless
Density 4.5 g/mL
Assay (metals basis)
Chemical Name or Material Barium sulfate
Show More Show Less
EINECSNumber 231-784-4
RTECSNumber CR0600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.