Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium sulfide, 99.7% (metals basis), Thermo Scientific Chemicals

Catalog Number AA1284809
Click to view available options
Quantity:
10 g
50 g
21109-95-5
BaS
169.39
MFCD00014192
CJDPJFRMHVXWPT-UHFFFAOYSA-N
barium sulfide, thioxobarium
88791
barium(2+) sulfanediide
[S--].[Ba++]
21109-95-5
BaS
169.39
MFCD00014192
CJDPJFRMHVXWPT-UHFFFAOYSA-N
barium sulfide, thioxobarium
88791
barium(2+) sulfanediide
[S--].[Ba++]

Barium sulfide has different characters, so it is used in a variety of ways, such as in electronics, paint pigments, dehairing hides, flame retardant, luminous paints, and generating pure hydrogen sulfide. Barium sulfide can deviate X-rays and helps to give clearly image of the soft tissue. Barium sulfide is a precursor to other barium compounds and also used as a short wavelength emitters for electronic displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1200°C
Color Gray to Yellow
Physical Form Powder
Quantity 10 g
UN Number UN3134
Merck Index 14,995
Solubility Information Soluble in water but insoluble in alcohol.
Formula Weight 169.4
Percent Purity 99.7%
Odor Like rotten eggs
Sensitivity Moisture sensitive
Density 4.25 g/mL
Assay (metals basis)
Chemical Name or Material Barium sulfide
Show More Show Less
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: III; Proper Shipping Name: WATER-REACTIVE SOLID, TOXIC, N.O.S.
EINECSNumber 244-214-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit