Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium titanate(IV), 99%, ACROS Organics™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Plastic bottle
12047-27-7
BaO3Ti
233.24
MFCD00003447
12047-27-7
BaO3Ti
233.24
MFCD00003447
CAS Min % 98.5
CAS Max % 100.0
Additional Information SrO: 0.15% max.
Assay Percent Range 0.988 to 1.000 (BaO+ SrO)/TiO2 Mole Ratio
Aluminum Oxide 0.04% max.
Linear Formula BaTiO3
Merck Index 15, 996
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in alkalies,slightly soluble in diluted acids,soluble in concentrated h2so4 and hf
Formula Weight 233.24
Physical Form Powder
Percent Purity 99%
Density 6.0800g/mL
Loss on Ignition 0.5% max.
Packaging Plastic bottle
Particle Size 0.03% Max. (+325 Mesh), 0.85 to 1.45 Micron (Horiba, D50)
Silicon Dioxide 0.02% max.
Sodium Oxide 0.03% max.
Solubility Solubility in water: insoluble. Other solubilities: insoluble in alkalies, slightly soluble in diluted acids, soluble in concentrated h2so4 and hf
Specific Gravity 6.08
Color White
Melting Point 1625.0°C
Quantity 500g
Chemical Name or Material Barium titanate(IV), 99%
Show More Show Less

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Warning

EINECSNumber : 234-975-0

RTECSNumber : XR1437333

TSCA : TSCA