Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium titanium oxide, 99.7% (metals basis)

Catalog No. p-7080442
Click to view available options
Quantity:
100 g
500 g

CAS: 12047-27-7 | BaO3Ti | 233.19 g/mol

Barium titanium oxide is a dielectric ceramic widely used in multi-layer ceramic capacitors (MLCCs), embedded decoupling capacitors (EDC) and ceramic filters. It is a piezoelectric and ferroelectric component used in microphones and transducers. It is also useful for thermistors and self-regulating electric heating systems. It is doped with small amounts of metals like scandium, yttrium, neodymium, samarium to become a semiconductor. It is used in some types of uncooled sensors for thermal cameras and in electric vehicles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12047-27-7
Molecular Formula BaO3Ti
Molecular Weight (g/mol) 233.19
MDL Number MFCD00003447
InChI Key WNKMTAQXMLAYHX-UHFFFAOYSA-N
Synonym Barium titanate
IUPAC Name barium(2+) oxotitaniumbis(olate)
SMILES [Ba++].[O-][Ti]([O-])=O

Specifications

Melting Point 1620°C
Color White
Physical Form Powder
Quantity 100 g
Merck Index 14,1000
Solubility Information Slightly soluble in dilute mineral acids. Soluble in concentrated hydrofluoric acid. Insoluble in water.
Formula Weight 233.19
Percent Purity 99.7%
Odor Odorless
Density 5.85 g/mL
Assay (metals basis)
Chemical Name or Material Barium titanium oxide
Show More Show Less
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 234-975-0
RTECSNumber XR1437333
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.