Alfa Aesar™ Barium titanium oxide, 99.95% (metals basis)

Manufacturer: Alfa Aesar™ 4508314

Quantity 25g

 View more versions of this product

Catalog No. AA4508314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12047-27-7
    Molecular Formula BaTiO3
    Formula Weight 233.19
    MDL Number MFCD00003447
    Merck Index 14,1000
    Synonym Barium titanate
    Chemical Name or Material Barium titanium oxide

    Specifications

    Formula Weight 233.19
    Health Hazard 3 GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Molecular Formula BaTiO3
    Health Hazard 2 GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.
    TSCA TSCA
    Particle Size 3 to 6mm
    Physical Form Pieces
    MDL Number MFCD00003447
    CAS 12047-27-7
    Quantity 25g
    Merck Index 14,1000
    Synonym Barium titanate
    Melting Point 1620°C
    Health Hazard 1 Warning
    Chemical Name or Material Barium titanium oxide
    Assay 99.95%, (metals basis)

    GHS H Statement
    H302-H332
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P261-P264-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wash thoroughly after handling.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit