Learn More
Barium titanium oxide, 99%
Description
Barium titanium oxide is a dielectric ceramic used for capacitors. It is a piezoelectric material for microphones and other transducers. Finds applications in nonlinear optics and photorefractive applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1620°C |
| Physical Form | Powder |
| Quantity | 500 g |
| Merck Index | 14,1000 |
| Solubility Information | Insoluble in water, alkalies; Slightly soluble in dilute acids; Soluble in concentrated acids |
| Formula Weight | 233.19 |
| Percent Purity | 99% |
| Odor | Odorless |
| Density | 5.85 g/mL |
| Assay | (metals basis) |
| Show More |
Safety and Handling
| Hazard Category | H302+H332 |
| Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c |
| EINECSNumber | 234-975-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.