Learn More
Thermo Scientific™ Barnstead™ Cartridge Kits
Click to view available options
For Use With (Equipment):
Nanopure II, Older Nanopure Systems
Nanopure Infinity Systems
Nanopure Quad-style System
Feedwater Source:
Deionized, Tap
Pretreated
Reverse Osmosis, Distilled
Tap
Tap or Pretreated
Model:
D3804
D3805
D3807
D4801
D4802
D5025
D50254
D50255
D50256
D5026
Description
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Specifications
Specifications
Feedwater Source | Reverse Osmosis, Distilled |
For Use With (Application) | Water purification cartridge |
For Use With (Equipment) | Nanopure Infinity Systems |
Includes | One 09-050-241 reverse osmosis/Distilled Feed Pretreatment, 09-050-246 Organicfree cartridge, two 09-050-247 Ultrapure™ cartridges and two 09-050-279 Remote Dispenser 0.2μm filters |
Model | D50255 |
Type | Cartridge Kit |