Learn More
Thermo Scientific™ Barnstead™ Cartridge Kits
GSA_VA
Catalog No. p-169245
Click to view available options
For Use With (Equipment):
Nanopure II, Older Nanopure Systems
Nanopure Infinity Systems
Nanopure Quad-style System
Feedwater Source:
Deionized, Tap
Pretreated
Reverse Osmosis, Distilled
Tap
Tap or Pretreated
Model:
D3804
D3805
D3807
D4801
D4802
D5025
D50254
D50255
D50256
D5026
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Feedwater Source Pretreated
For Use With (Application) Water purification cartridge
For Use With (Equipment) Nanopure Quad-style System
Includes One 09-034-5C Macropure™ Pretreatment, 09-050-267 Organicfree, 09-050-276 Ultrapure™ and FS090343 Ultrapure™ cartridges
Model D5026
Type Cartridge Kit
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.