Learn More
Thermo Scientific™ Barnstead™ Classic Still Storage Accessories
Catalog No. p-4531439
Click to view available options
Type:
Air Filter and Water Seal
Catalog No. 09-050-115
Supplier: Thermo Scientific™ H3111
Only null left
Add to Cart
Add to Cart
Complete your distillation system with Thermo Scientific™ Barnstead™ Classic Still Storage Accessories.
Thermo Scientific™ Barnstead™ Classic Still Storage Accessories complement your current system to create a more complete system. Accessories include mounting options, UV light assembly, Thermo Scientific™ Ventgard™ air filter and water seal. Choose either a floor stand or wall bracket as needed. Maintain sterility in the reservoir with the UV light assembly, which includes housing, on/off switch, 6' cord, lamp and ballast. Protect stored water against airborne particulates, organics and CO2 with Ventgard™ air filter and water seal. Prevent air from entering the reservoir via the condenser atmospheric vent.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Classic Reservoir
Type Air Filter and Water Seal
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.