Learn More
Thermo Scientific™ Barnstead™ Classic Stills
Description
Remove inorganic solids, organics, bacteria and pyrogens from your water through the distillation process using Thermo Scientific™ Barnstead™ Classic Stills. Choose from bench or floor mounted models. Available in five capacities to meet your production needs, stills are constructed of copper and bronze with a pure tin coating. The inert nature of tin prevents leaching of contaminants into water. Portable stills allow for quick set-up, while floor model electric stills improve safety with over-temperature protection. Steam stills feature optional automatic controls for unattended operation.
- Double-walled boiler and preheating of feed water conserves electricity.
- Vented condenser allows for stripping of gaseous impurities.
- Unique de-concentrator removes scale forming impurities from the boiler.
- Unique Q-Baffle ensures high-quality pyrogen-free water by stripping contaminant laden water droplets from steam.
- Still includes space-saving horizontal condenser.
- Metal construction withstands years of use.
- Inert pure tin pathways assure product water quality.
Portable Stills:
- Require no permanent plumbing or electrical connections allowing for quick setup
- Include low water cutoff for safety protection
- Bench mounted
Floor Model Electric Still:
- Comes standard with floor mounting stand
- Improves safety with new overtemperature protection system
- Allows for easier installation with new plug-and-play hardware
- Processes capacities from 1 to 10 gal./hr.
- Includes low water cutoff that protects heating elements when water supply is interrupted
- Allows for unattended operation with optional fully automatic controls
- Stops still when storage tank is full, starts still when water level falls below preset level
- Drains still boiler at selected two-, four-, eight- and 16-hour intervals to reduce scale formation
Steam Still:
- Utilizes available in-house steam as heat source
- Optional fully automatic controls allow for unattended operation
- Stops still when storage tank is full, starts still when water level falls below preset level
- Drains still boiler at selected two-, four-, eight- and 16-hour intervals to reduce scale formation
- Steam pressure: 35 to 50psi
- Boiler horsepower: 3.5
- Steam flowrate: 90 lb./hr. (41kg/hr.)
Ordering Information:
Automated control systems are available for all stills: G2100 (for 1 gal./hr., 2 gal./hr. models), G2110 (5 gal./hr.), G2125 (10 gal./hr.) and G2010 (10 gal./hr.). Order separately.
This product is not available in all regions.
Includes:
Floor stand (exept for Portable Model A1007).
Requires:
Storage reservoir, UV light assembly and Ventgard Air Filter (order separately)
Recommended for:
Buffer/reagent prep, glassware washing, dishwasher feed

Specifications
Specifications
Type | Electric |
Amperage | 26 A |
Voltage | 240 V |
Phase | 1 |
Frequency | 50/60 Hz |
Output Flow Rate | 2 gal./hr. |
Electrical Requirements | 240 V 50/60 Hz |
Dimensions (L x W x H) | 11 x 23 x 68 in. (28 x 58 x 173 cm) |
Flow Rate | 2 gal./hr. |
Model | Floor Electric with Stand |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.