Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Barnstead™ Classic Stills

Catalog No. 09720343
Click to view available options
Output Flow Rate:
0.5 gal./hr.
10 gal./hr.
2 gal./hr.
5 gal./hr.
Type:
Electric
Electric (Portable)
Electrical Requirements:
120 V 50/60 Hz
208 V 50/60 Hz
240 V 50/60 Hz
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Remove inorganic solids, organics, bacteria and pyrogens from your water with Thermo Scientific™ Barnstead™ Classic Stills.

Remove inorganic solids, organics, bacteria and pyrogens from your water through the distillation process using Thermo Scientific™ Barnstead™ Classic Stills. Choose from bench or floor mounted models. Available in five capacities to meet your production needs, stills are constructed of copper and bronze with a pure tin coating. The inert nature of tin prevents leaching of contaminants into water. Portable stills allow for quick set-up, while floor model electric stills improve safety with over-temperature protection. Steam stills feature optional automatic controls for unattended operation.

  • Double-walled boiler and preheating of feed water conserves electricity.
  • Vented condenser allows for stripping of gaseous impurities.
  • Unique de-concentrator removes scale forming impurities from the boiler.
  • Unique Q-Baffle ensures high-quality pyrogen-free water by stripping contaminant laden water droplets from steam.
  • Still includes space-saving horizontal condenser.
  • Metal construction withstands years of use.
  • Inert pure tin pathways assure product water quality.

Portable Stills:

  • Require no permanent plumbing or electrical connections allowing for quick setup
  • Include low water cutoff for safety protection
  • Bench mounted

Floor Model Electric Still:

  • Comes standard with floor mounting stand
  • Improves safety with new overtemperature protection system
  • Allows for easier installation with new plug-and-play hardware
  • Processes capacities from 1 to 10 gal./hr.
  • Includes low water cutoff that protects heating elements when water supply is interrupted
  • Allows for unattended operation with optional fully automatic controls
  • Stops still when storage tank is full, starts still when water level falls below preset level
  • Drains still boiler at selected two-, four-, eight- and 16-hour intervals to reduce scale formation

Steam Still:

  • Utilizes available in-house steam as heat source
  • Optional fully automatic controls allow for unattended operation
  • Stops still when storage tank is full, starts still when water level falls below preset level
  • Drains still boiler at selected two-, four-, eight- and 16-hour intervals to reduce scale formation
  • Steam pressure: 35 to 50psi
  • Boiler horsepower: 3.5
  • Steam flowrate: 90 lb./hr. (41kg/hr.)

Ordering Information:

Automated control systems are available for all stills: G2100 (for 1 gal./hr., 2 gal./hr. models), G2110 (5 gal./hr.), G2125 (10 gal./hr.) and G2010 (10 gal./hr.). Order separately.

This product is not available in all regions.

Includes:

Floor stand (exept for Portable Model A1007).

Requires:

Storage reservoir, UV light assembly and Ventgard Air Filter (order separately)

Recommended for:

Buffer/reagent prep, glassware washing, dishwasher feed

TRUSTED_SUSTAINABILITY

Specifications

Type Electric
Voltage 240 V
Frequency 50/60 Hz
Phase 1
Amperage 57 A
Output Flow Rate 5 gal./hr.
Electrical Requirements 240 V 50/60 Hz
Flow Rate 5 gal./hr.
Dimensions (L x W x H) 14 x 35 x 77 in. (36 x 89 x 196 cm)
Model Floor Electric with Stand
Height (English) 77 in.
Height (Metric) 196 cm
Width (English) 35 in.
Width (Metric) 89 cm
Cooling Flow Rate (Metric) 151 L/hr.
Length (English) 14 in.
Length (Metric) 36 cm
Certifications/Compliance UL, cUL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.