Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories
GSA_VA
Catalog No. p-4531495
Change view
Click to view available options
Type:
Remote Dispenser
Voltage:
0/1 V
Catalog No. Type Voltage
09-035-34 Remote Dispenser 0/1 V
1 options
Catalog No. 09-035-34 Supplier Thermo Scientific™ Supplier No. D8952
Only null left
Add to Cart
Add to Cart
Optimize your ultrapure water purification system with Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories, including a remote dispenser.
Optimize your ultrapure water purification system with Thermo Scientific™ Barnstead™ E-Pure™ Optional Accessories. Increase flexibility in your lab with the remote dispenser, which delivers water up to 8' (2.4m) from the system and ships with a mounting bracket. Gain protection with the low pressure pump protector which alerts the E-Pure of an inadequate feed water condition in a pressurized feed line. The low water level pump protector alerts the system of an inadequate feed water condition when fed from a reservoir.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Barnstead E-Pure
Type Remote Dispenser
Voltage 0/1 V