Learn More
Thermo Scientific™ Barnstead™ E-Pure™ Ultrapure Water Purification Systems

Description
Produce Type 1 ultrapure water with low operating costs using Thermo Scientific™ Barnstead™ E-Pure™ Ultrapure Water Purification Systems. Choose from a three- or four-module system. Gain longer cartridge life and benefit from lower operating costs with large cartridges. This system is Ideal for AA, GS, buffer preps, or as a purified feed water source for analytical instruments or labware washers. It is simple to use and easy to maintain.
AquaPro™ digital on-line analytical measurement platform featuring:
- Large color graphic display
- Temperature readout
- Temperature compensated
- Calibratable
- System achieves resistivities up to 18.2 MΩ-cm and TOC levels below 10ppb.
- Virgin polypropylene water pathways prevent recontamination.
- Recirculation pump continuously recirculates water throughout the entire system to maintain water purity and reduce the quantity of wastewater required for rinse-up.
- System is wall-mountable and completely assembled for easy installation.
- Quarter-turn quick release canisters allow for easy cartridge replacement.
- 0.2μm absolute final filter removes bacteria and particulates that could alter your results.
- Digital resistivity meter is automatically temperature-compensated to 25°C, preventing fluctuating readings due to temperature changes.
Ordering Information:
Optional accessories include Remote Dispenser (D8952), Low Pressure Pump Protector (D2706), and Low Water Level Pump Protector (D0603, D0606). These products, as well as replacement cartridges and consumables, are available separately. For details, contact your sales representative.
Includes:
Wall bracket
Requires:
O-rings included with cartridge pack (sold separately)
Recommended for:
AA, GS, buffer preps, feedwater for analytical instruments or labware washers
Note:
If feeding E-Pure from a storage reservoir, Low Water Pump Protector is recommended to protect pump from damage if reservoir water level falls below system requirements. Order separately (D0603 for 120V, D0606 for 240V).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Certifications/Compliance | CE, CSA |
Conductivity | Type 1: 0.055μS/cm, Type 2: 0.067 to 0.1μS/cm |
Bacterial Content | <1 CFU/mL |
Feed Water Monitoring | No |
Feedwater Source | Tap or Pretreated |
Flow Rate | Up to 2.5 L/min. |
Product Water Type | Type 1 |
Remote Dispenser | Optional |
Resistivity | 18.2 Mohm.cm at 25°C |
TOC Monitor with UV Intensity Monitoring | No |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.