Learn More
Description
Disposable nitrile gloves with TNT™ chemical splash resistance technology offer a high level of protection against harmful exposures.
- 2X more chemical splash protection than leading nitrile brands
- High standard for barrier quality and consistency (Low 0.40 AQL)
- Robust construction prevents rips, tears and breaches
- Made with proprietary TNT™ Technology for soft, durable protection against a wide range of chemicals
- Silicone-free formulation for enhanced product protection
Specifications
Specifications
| Certifications/Compliance | ASTM D6319, FDA 21 CFR 177.2600, ASTM F1671 |
| Certifications/Compliance (Europe) | Category III, EN 420:2003 + A1:2009, EN ISO 374-1:2016 |
| Chlorinated | No |
| AQL | 0.4 |
| Color | Green |
| Disposable | Single-use |
| Finger Thickness (English) | 7.5 mil |
| Finger Thickness (Metric) | 0.19 mm |
| Hand | Ambidextrous |
| Latex Free | Yes |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.