Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BarX1 Antibody (BARX1/2760), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308671AF700
Description
BarX1 Monoclonal antibody specifically detects BarX1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
BarX1 | |
Monoclonal | |
Alexa Fluor 700 | |
BarH-like homeobox 1, BARX homeobox 1, homeobox protein BarH-like 1 | |
Recombinant full-length human BarX1 protein. (Uniprot: Q9HU1) | |
100 μg | |
Primary | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
BARX1/2760 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
56033 | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction