Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BarX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565984
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565984 Supplier Invitrogen™ Supplier No. PA565984
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSIPTSEQLTE QERAKDAEKP AEVPGEPSDR SRED Highest antigen sequence identity to the following orthologs - mouse 86%, rat 91%.

BarX1 is a member of the Bar subclass of homeobox transcription factors. Studies of the mouse and chick homolog suggest the encoded protein may play a role in developing teeth and craniofacial mesenchyme of neural crest origin. The protein may also be associated with differentiation of stomach epithelia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BarX1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BARX1
Gene Accession No. Q9HBU1
Gene Alias BarH-like homeobox 1; BARX homeobox 1; BARX1; Homeobox protein BarH-like 1; LOW QUALITY PROTEIN: homeobox protein BarH-like 1
Gene Symbols BARX1
Host Species Rabbit
Immunogen Recombinant Human BarX1. Recombinant protein control fragment (Product #RP-106671).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56033
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.