Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BarX2 Monoclonal Antibody (BAR X2 8A7/1)
GREENER_CHOICE

Catalog No. PIMA127164
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA127164 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA127164 Supplier Invitrogen™ Supplier No. MA127164
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sections need microwave pre-treatment. A molecular weight of approximately 31 kDa is predicted.

Transcription factor. Binds optimally to the DNA consensus sequence 5'-YYTAATGRTTTTY-3'. May control the expression of neural adhesion molecules such as L1 or Ng-CAM during embryonic development of both the central and peripherical nervous system. May be involved in controlling adhesive processes in keratinizing epithelia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BarX2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone BAR X2 8A7/1
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene BARX2
Gene Accession No. O08686, Q9UMQ3
Gene Alias 2310006E12Rik; BarH-like homeobox 2; BARX homeobox 2; BARX homeobox 2, pseudogene 1; BARX2; Barx2b; Barx2-ps1; homeobox protein BarH-like 2; homeodomain transcription factor BARX2
Gene Symbols BARX2
Host Species Mouse
Immunogen Fusion protein: MBP-tagged BarX2.
Purification Method IgG fraction
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12023, 8538
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.