Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BarX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566991
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566991 Supplier Invitrogen™ Supplier No. PA566991
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSEEIEAEEKM NSQAQGQEQL EPSQGQEELC EAQEPKARDV PLEMAEPPDP PQELPIPSS Highest antigen sequence identity to the following orthologs - mouse 72%, rat 73%.

Transcription factor. Binds optimally to the DNA consensus sequence 5'-YYTAATGRTTTTY-3'. May control the expression of neural adhesion molecules such as L1 or Ng-CAM during embryonic development of both the central and peripherical nervous system. May be involved in controlling adhesive processes in keratinizing epithelia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BarX2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BARX2
Gene Accession No. Q9UMQ3
Gene Alias 2310006E12Rik; BarH-like homeobox 2; BARX homeobox 2; BARX homeobox 2, pseudogene 1; BARX2; Barx2b; Barx2-ps1; homeobox protein BarH-like 2; homeodomain transcription factor BARX2
Gene Symbols BARX2
Host Species Rabbit
Immunogen Recombinant Human BarX2. Recombinant protein control fragment (Product #RP-107636).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8538
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.