Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BarX2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984302
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4302 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4302 Supplier Bioss Supplier No. BS19273R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transcription factor. Binds optimally to the DNA consensus sequence 5′-YYTAATGRTTTTY-3′. May control the expression of neural adhesion molecules such as L1 or Ng-CAM during embryonic development of both the central and peripherical nervous system. May be involved in controlling adhesive processes in keratinizing epithelia.

Specifications

Antigen BarX2
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BARX2
Gene Accession No. Q9UMQ3
Gene Alias 2310006E12Rik; BarH-like homeobox 2; BARX homeobox 2; Barx2; Barx2b; Homeobox protein BarH-like 2; homeodomain transcription factor BARX2
Gene Symbols BARX2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BarX2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8538
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.