Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Basal Bodies of Cillia Antibody (LhS 28), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250114MFV450
Description
Basal Bodies of Cillia Monoclonal antibody specifically detects Basal Bodies of Cillia in Human, Hamster samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| Basal Bodies of Cillia | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| BHK a21 Cytoskeletal preparation | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunocytochemistry, Immunoprecipitation | |
| LhS 28 | |
| Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human, Hamster | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction