Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Basal cell cytokeratin Antibody (RCK103), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197729PE
Description
Basal cell cytokeratin Monoclonal specifically detects Basal cell cytokeratin in Human, Rat, Porcine, Avian, Canine, Chicken, Guinea Pig, Hamster, Rabbit, Zebrafish samples. It is validated for Flow Cytometry.Specifications
Basal cell cytokeratin | |
Monoclonal | |
PE | |
Mouse | |
Affinity Purified | |
RUO | |
This monoclonal antibody stains basal cells in combined and stratified epithelial tissues. It recognizes the stem cell population, including the so-called amplifying cells in the prostate epithelium. | |
Store at 4C in the dark. |
Flow Cytometry | |
RCK103 | |
Flow Cytometry | |
RCK103 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a mix of cell preparations containing Human Cytokeratins. | |
0.1 mL | |
Primary | |
Human, Rat, Pig, Avian, Canine, Chicken, Guinea Pig, Hamster, Rabbit, Zebrafish | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction