Description
- Aphanitic volcanic rock forms from effusive eruptions, such as shield volcanoes of Hawaii, or deep ocean floor
- For use in introduction to igneous rock classification and identification
- Correlates well to Next Generation Science Standards Evidence Statements
- Students can recreate cooling history of this sample based on texture and abundance of phenocrysts
- Example of aphanitic igneous rock
Specifications
Specifications
| Product Type | Igneous Rock |
| For Use With (Application) | Introduction to igneous rock classification and identification |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 12/Pk. |
| Specimen Size | 2 to 3 cm |
| Specimen Type | Basalt |