Learn More
Corning™ Base Head Clamp
GSA_VAGREENER_CHOICE
Catalog No. 07250147 Shop All Corning Products
Change view
Click to view available options
For Use With (Equipment):
Holding Rod
Catalog No. For Use With (Equipment)
07-250-147 Holding Rod
1 options
This item is not returnable. View return policy
Catalog No. 07-250-147 Supplier Corning™ Supplier No. 440140
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
For Use With: Holding Rod
Corning offers a 2 diameter support rods. It is designed to be used with the Corning Temperature probe support clamp (Cat. No. 440141).
Note: Bosshead clamps to support rod; holding rod connects to bosshead. Temperature controller slides into holding rod.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Holding Rod
Type Clamp
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov