Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Basic Agarose Premier
Click to view available options
Quantity:
100 g
500 g
Description
- Agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae.
- Basic agarose premier is ideal for routine rapid separation of DNA and RNA fragments as well as PCR products, the preparation of plasmids, and for screening, cloning and blotting techniques.

Specifications
Specifications
CAS | 9012-36-6 |
Description | Agarose is a purified lineargalactan hydrocolloid isolated from agar or agar-bearing marine algae. |
Color | White |
Product Type | Agarose |
Form | Powder |
For Use With (Application) | Analysis of nucleic acids bygel electrophoresis |
Quantity | 500 g |
Storage Requirements | Store at Room Temperature (15-30°C). |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction