Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Basic Fuchsin, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Basic Fuchsin is a magenta dye that is a combination of rosanilin hydrochloride, pararosaniline, new fuchsine and magenta II. Basic Fuchsin is a dark green crystal often used in the laboratory where it is employed to stain bacteria and in the fabrication of Schiff reagent. It is made up of a combination of rosanilin hydrochloride, pararosaniline, new fuchsine and magenta II. Basic Fuchsin also has uses in the textile industry, and has many other names, including Basic Parafuchsin, Basic Red 9, Paramagenta Hydrochloride, and Pararosaniline Chloride.
Specifications
Specifications
CAS Max % | 100% |
Packaging | Amber Glass Bottle |
Quantity | 25 g |
Grade | Reagent |
Physical Form | Crystalline Powder or Lumps |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction