Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Basic Rocks and Minerals Kit
SCI_ED

Catalog No. s24663 Shop All Eisco Products
Change view
Click to view available options
:
Basic Rocks and Minerals Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S24663 Basic Rocks and Minerals Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24663 Supplier Eisco™ Supplier No. ESNG0004

For use in hands-on exploration of rocks and minerals in classroom setting. The Basic Rocks and Minerals Kit contains 40 specimens and specimen key.

  • For use in learning how to identify rocks and minerals outdoors
  • Added collection for first-time and experienced rock hounds
  • Average specimen size: 2 to 3 cm
  • Offers comprehensive overview of rocks and minerals
  • Contains rocks from inside of volcanoes, bottom of sea floor, from deep within earth's crust and more

Specifications

Product Type Basic Rocks and Minerals Kit
For Use With (Application) Identify/compare geologic samples
Includes Pyrite, Galena, Hematite, Magnetite, Limonite, Graphite, Calcite, Gypsum, Halite, Fluorite, Garnet, Hornblende, Biotite, Muscovite, Microcline, Albite, Olivite, Milky Quartz, Sulfur, Talc, Basalt, Gabbro, Porphyritic Granite, Obsidian, Pumice, Phyolite, Plagioclase, B, Granite, Basalt, Conglomerate, Limestone, Gneiss, and Marble
Packaging Sectioned container
Quantity 40 Specimens
Specimen Size Approx. 1 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.