Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Basophil Marker Monoclonal Antibody (2D7)
GREENER_CHOICE

Catalog No. FERMA548986
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548986 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548986 Supplier Invitrogen™ Supplier No. MA548986
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody 2D7 is a specific marker for basophils detecting a basophil-specific protein of 7.2-7.5 kDa localized in secretory granules. The 2D7 antibody can be used as a sensitive and precise marker for human basophils and does not react with lymphocytes, monocytes, eosinophils, neutrophils or mast cells in immunohistochemistry.

The human 2D7 antigen is expressed exclusively by human basophils. This human basophil marker is a 7.2-7.5 kDa protein present exclusively in the secretory granules of human basophils. Upon activation, basophils may degranulate, thereby decreasing the amount of 2D7 antigen available for staining.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Basophil Marker
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2D7
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene Alias Basophil 2D7 marker; Basophils
Host Species Mouse
Immunogen Lysate of purified basophils.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.