Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Basophils (2D7) Mouse anti-Human, R718, Clone: 2D7/BASO (also known as 2D7), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 568380
This item is not returnable.
View return policy
Description
Basophils (2D7) Monoclonal antibody specifically detects Basophils (2D7) in Human samples. It is validated for Intracellular staining (flow cytometry) (Routinely Tested)
Specifications
| Basophils (2D7) | |
| Monoclonal | |
| R718 | |
| Basophils; Basophils (2D7); 2D7 Antigen; 2D7 Ligand | |
| Purified Human Basophil Lysate | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. | |
| IgG1 κ |
| Flow Cytometry | |
| 2D7/BASO (also known as 2D7) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction