Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Basophils Antibody (2D7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311212
Description
Basophils Monoclonal specifically detects Basophils in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Basophils | |
Monoclonal | |
0.1 mg/ml | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Anti-Basophils, Basophil | |
Lysate of purified basophils | |
50 μg | |
Primary | |
Store at 4°C. Do not freeze. | |
IgG1 |
Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
2D7 | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction