Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BASP1 Antibody, Novus Biologicals™
SDP

Catalog No. p-7108627 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP168985 100 μL
NBP16898520 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP168985 Supplier Novus Biologicals Supplier No. NBP168985
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BASP1 Polyclonal specifically detects BASP1 in Human, Rat samples. It is validated for Western Blot.

Specifications

Antigen BASP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P80723
Gene Alias brain abundant, membrane attached signal protein 1,22 kDa neuronal tissue-enriched acidic protein, brain acid-soluble protein 1, CAP-23, CAP23brain acid soluble protein 1, NAP-22, NAP22MGC8555, Neuronal axonal membrane protein NAP-22, neuronal tissue-enriched acidic protein
Gene Symbols BASP1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to BASP1 (brain abundant, membrane attached signal protein 1) The peptide sequence was selected from the C terminal of BASP1. Peptide sequence APAAGEEPSKEEGEPKKTEAPAAPAAQETKSDGAPASDSKPGSSEAAPSS.
Molecular Weight of Antigen 23 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 10409
Test Specificity Yeast: 82%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Rat, Rabbit, Yeast
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.