Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BASP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114894 Supplier Invitrogen™ Supplier No. PA5114894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BASP1.

BASP1 (brain acid soluble protein 1) is a membrane bound protein with several transient phosphorylation sites and PEST motifs. Conservation of proteins with PEST sequences among different species supports their functional significance. PEST sequences typically occur in proteins with high turnover rates. Immunological characteristics of this protein are species specific. This protein also undergoes N-terminal myristoylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BASP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene BASP1
Gene Accession No. P80723, Q05175, Q91XV3
Gene Alias 22 kDa neuronal tissue-enriched acidic protein; 2610024P12Rik; BASP1; brain abundant membrane attached signal protein 1; brain abundant, membrane attached signal protein 1; brain acid soluble protein 1; brain acidic membrane protein; brain acid-soluble protein 1; CAP23; CAP-23; Ckap3; cytoskeleton associated protein 3; MGC8555; NAP22; NAP-22; neuronal axonal membrane protein NAP-22; neuronal tissue-enriched acidic protein
Gene Symbols BASP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human BASP1(Accession P80723), corresponding to amino acid residues M1-E39.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10409, 64160, 70350
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.