Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BASP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA526969
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA526969 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA526969 Supplier Invitrogen™ Supplier No. PA526969
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BASP1 Polyclonal Antibody for Western Blot

BASP1 (brain acid soluble protein 1) is a membrane bound protein with several transient phosphorylation sites and PEST motifs. Conservation of proteins with PEST sequences among different species supports their functional significance. PEST sequences typically occur in proteins with high turnover rates. Immunological characteristics of this protein are species specific. This protein also undergoes N-terminal myristoylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BASP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene BASP1
Gene Accession No. P80723
Gene Alias 22 kDa neuronal tissue-enriched acidic protein; 2610024P12Rik; BASP1; brain abundant membrane attached signal protein 1; brain abundant, membrane attached signal protein 1; brain acid soluble protein 1; brain acidic membrane protein; brain acid-soluble protein 1; CAP23; CAP-23; Ckap3; cytoskeleton associated protein 3; MGC8555; NAP22; NAP-22; neuronal axonal membrane protein NAP-22; neuronal tissue-enriched acidic protein
Gene Symbols BASP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 123-150 amino acids from the central region of human BASP1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10409
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.