Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bassoon Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502278204
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8204 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8204 Supplier Leinco Technologies Inc Supplier No. 56495100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Bassoon Polyclonal antibody specifically detects Bassoon in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blot

Specifications

Antigen Bassoon
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene BSN
Gene Accession No. O88737, O88778, Q9UPA5
Gene Alias bassoon; bassoon (presynaptic cytomatrix protein); bassoon presynaptic cytomatrix protein; BSN; Kiaa0434; neuronal double zinc finger protein; presynaptic cytomatrix protein; protein bassoon; Zinc finger protein 231; ZNF231
Host Species Rabbit
Immunogen Fusion protein corresponding to aa786-1041 (N-terminal) of human Bassoon (accession no. NM_003458.3).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12217, 29138, 8927
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.