Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bat Immunoglobulin Antibody (BT1-4F10), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP223483AF700
Description
Bat Immunoglobulin Monoclonal specifically detects Bat Immunoglobulin in Bat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Bat Immunoglobulin | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Microchiropteran bat (Eptesicus fuscus) spleen cells. | |
0.1 mL | |
Primary | |
Bat | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
BT1-4F10 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein G purified | |
RUO | |
Immunoglobulin L (Light) chain. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction