Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bat Immunoglobulin Antibody (BT1-4F10), R&D Systems™
SDP

Catalog No. p-4964336 Shop All Novus Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223483SS 0.025 mL
NBP223483 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP223483SS Supplier R&D Systems Supplier No. NBP223483SS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody has been used in 2 publications

Bat Immunoglobulin Monoclonal specifically detects Bat Immunoglobulin in Bat samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot.

Specifications

Antigen Bat Immunoglobulin
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone BT1-4F10
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml-1.0 ug/ml, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation
Host Species Mouse
Immunogen Microchiropteran bat (Eptesicus fuscus) spleen cells.
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Immunoglobulin L (Light) chain.
Target Species Bat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.