Learn More
BAT3 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302039AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
BAT3 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P46379 | |
BAG6 | |
Between 1082 and 1132 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
BAG6 | |
DAQB-195H10.3, BAG-6, BAG6, D6S52E, G3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7917 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.