Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BAT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563596
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563596 Supplier Invitrogen™ Supplier No. PA563596
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLVTRTVRQH LNLNNAEQLC RYQGPVLLIR RTKDEIITTT VPEDIM Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.

A cluster of genes, BAT1-BAT5, has been localized in the vicinity of the genes for TNF alpha and TNF beta. These genes are all within the human major histocompatibility complex class III region. The protein encoded by this gene is thought to be involved in some aspects of immunity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BAT5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ABHD16A
Gene Accession No. O95870
Gene Alias Abhd16a; abhydrolase domain containing 16A; abhydrolase domain-containing protein 16A; AI326074; Alpha/beta hydrolase domain-containing protein 16A; Bat5; Bat-5; D17H6S82E; D6S82E; G5; hBAT5; HLA-B associated transcript 5; HLA-B-associated transcript 5; HLA-B-associated transcript 5 homolog; mBAT5; Monoacylglycerol lipase ABHD16A; Ng26; Phosphatidylserine lipase ABHD16A; PP199; protein ABHD16A; protein BAT5; protein G5
Gene Symbols ABHD16A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BAT5. Recombinant protein control fragment (Product #RP-101614).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7920
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.