Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    BATF Antibody (687706), mFluor Violet 500 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8054MFV500
Description
BATF Monoclonal antibody specifically detects BATF in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| BATF | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
| Western Blot, Flow Cytometry, CyTOF | |
| 687706 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein | |
| E. coli-derived recombinant human BATF , Met1-Pro125, Accession # Q16520 | |
| 0.1 mL | |
| Signal Transduction | |
| 10538 | |
| Store at 4C in the dark. | |
| IgG2b | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            