Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BATF Antibody (687706), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8054P
Description
BATF Monoclonal antibody specifically detects BATF in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
BATF | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
687706 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein | |
E. coli-derived recombinant human BATF , Met1-Pro125, Accession # Q16520 | |
0.1 mL | |
Signal Transduction | |
10538 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction