Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BATF Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | BATF |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
BATF Polyclonal specifically detects BATF in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
BATF | |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
10538 | |
Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human BATF. The immunogen is located within the first 50 amino acids of BATF. Amino Acid Squence: SFSRSPPPGKQDSSDD | |
Primary | |
Multiple isoforms of BATF are known to exist. Despite its predicted molecular weight, BATF often migrates at a higher than expected molecular weight. |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
basic leucine zipper transcription factor, ATF-like, BATF1basic leucine zipper transcriptional factor ATF-like, B-ATFSFA2, B-cell-activating transcription factor, SFA-2activating transcription factor B, SF-HT-activated gene 2 protein | |
BATF | |
IgG | |
Affinity Purified | |
14 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title