Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BATF Recombinant Rabbit Monoclonal Antibody (HL1937)
GREENER_CHOICE

Catalog No. PIMA547227
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547227 Supplier Invitrogen™ Supplier No. MA547227
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

BATF is a nuclear basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. The leucine zipper of this protein mediates dimerization with members of the Jun family of proteins. This protein is thought to be a negative regulator of AP-1/ATF transcriptional events and blocks cellular transformation by Ras and Fos. BATF also is required for the differentiation of IL-17-producing T helper (TH17) cells, a CD4+ subset of T cells that coordinates the inflammatory response in host defense. In both T cells and B cells, BATF is required for the appropriate regulation of the class-switch recombination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BATF
Applications Western Blot
Classification Recombinant Monoclonal
Clone HL1937
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Batf
Gene Accession No. Q16520
Gene Alias Activating transcription factor B; basic leucine zipper ATF-like transcription factor; basic leucine zipper transcription factor, ATF-like; basic leucine zipper transcription factor-like; basic leucine zipper transcriptional factor ATF-like; BATF; B-ATF; BATF1; B-cell Activating transcription factor; B-cell-Activating transcription factor; SFA2; SFA-2; SF-HT-activated gene 2 protein; si:ch211-153j24.4
Gene Symbols Batf
Host Species Rabbit
Immunogen Recombinant fragment of human BATF.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10538
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.