Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BATF Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987785
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7785 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7785 Supplier Bioss Supplier No. BS7463R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BATF is a nuclear basic leucine zipper protein that belongs to the AP-1/ATF superfamily of transcription factors. The leucine zipper of this protein mediates dimerization with members of the Jun family of proteins. This protein is thought to be a negative regulator of AP-1/ATF transcriptional events and blocks cellular transformation by Ras and Fos. BATF also is required for the differentiation of IL-17-producing T helper (TH17) cells, a CD4+ subset of T cells that coordinates the inflammatory response in host defense. In both T cells and B cells, BATF is required for the appropriate regulation of the class-switch recombination.

Specifications

Antigen BATF
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Batf
Gene Accession No. Q16520
Gene Alias activating transcription factor B; basic leucine zipper ATF-like transcription factor; basic leucine zipper transcription factor, ATF-like; basic leucine zipper transcription factor-like; Basic leucine zipper transcriptional factor ATF-like; BATF; B-ATF; BATF1; B-cell activating transcription factor; B-cell-activating transcription factor; BOS_11116; LOC100338089; SFA2; SFA-2; SF-HT-activated gene 2 protein; si:ch211-153j24.4
Gene Symbols Batf
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BATF.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10538
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.