Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BATF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103647
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103647 Supplier Invitrogen™ Supplier No. PA5103647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BATF2.

BATF2 (basic leucine zipper transcription factor, ATF-like 2) is a 274 amino acid protein that localizes to the nucleus and contains one bZIP domain, suggesting that it may be involved in transcriptional regulation. The gene encoding BATF2, which is expressed as multiple alternatively spliced isoforms, is located on human chromosome 11. With approximately 135 million base pairs and 1, 400 genes, chromosome 11 comprises approximately 4% of human genomic DNA and is considered a gene and disease association dense chromosome. The chromosome 11 encoded Atm gene is important for regulation of cell cycle arrest and apoptosis following double strand DNA breaks. Atm mutation leads to the disorder known as ataxia-telangiectasia. The blood disorders Sickle cell anemia and thalassemia are caused by HBB gene mutations, while Wilms' tumors, WAGR syndrome and Denys-Drash syndrome are associated with mutations of the WT1 gene. Jervell and Lange-Nielsen syndrome, Jacobsen syndrome, Niemann-Pick disease, hereditary angioedema and Smith-Lemli-Opitz syndrome are also associated with defects in chromosome 11-encoded genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BATF2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BATF2
Gene Accession No. Q8N1L9
Gene Alias 4933430F08Rik; basic leucine zipper ATF-like transcription factor 2; basic leucine zipper transcription factor, ATF-like 2; basic leucine zipper transcriptional factor ATF-like 2; BATF2; B-ATF-2; MGC20410; RGD1311481; SARI; Suppressor of AP-1 regulated by IFN
Gene Symbols BATF2
Host Species Rabbit
Immunogen A synthesized peptide derived from human BATF2(Accession Q8N1L9), corresponding to amino acid residues S189-E239.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116071
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.