Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bathocuproin, 98%

Catalog No. AAB2284103
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
AAB2284103 1 g
AAB2284106 5 g
2 options

Catalog No. AAB2284103

Supplier: Thermo Scientific Chemicals B2284103

Only null left
Add to Cart

CAS: 4733-39-5 | C26H20N2 | 360.46 g/mol

It is used as a reagent for the determination of copper. It acts as an exciton blocking barrier which prohibits excitons diffusion process towards the Al electrode otherwise being quenched. It is the most commonly used buffer layer between acceptor and cathode layer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent for the determination of copper. It acts as an exciton blocking barrier which prohibits excitons diffusion process towards the Al electrode otherwise being quenched. It is the most commonly used buffer layer between acceptor and cathode layer.

Solubility
Soluble in methanol (36mg/100ml), ethanol, 2-propanol, bezene, and acetone. Partly miscible in water.

Notes
Store in cool, dry conditions in a sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4733-39-5
Molecular Formula C26H20N2
Molecular Weight (g/mol) 360.46
MDL Number MFCD00004972
InChI Key STTGYIUESPWXOW-UHFFFAOYSA-N
Synonym bathocuproine, bathocuproin, 1,10-phenanthroline, 2,9-dimethyl-4,7-diphenyl, unii-9thp2v94fx, 9thp2v94fx, 2,9-dimethyl-4,7-diphenylpyridino 3,2-h quinoline, pubchem19657, bathocuproin 1g, bidd:gt0563
PubChem CID 65149
IUPAC Name 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline
SMILES CC1=CC(C2=CC=CC=C2)=C2C=CC3=C(C=C(C)N=C3C2=N1)C1=CC=CC=C1

Specifications

Melting Point 279°C to 285°C
Quantity 1 g
Beilstein 306714
Solubility Information Soluble in methanol (36mg/100ml),ethanol,2-propanol,bezene,and acetone. Partly miscible in water.
Formula Weight 360.46
Percent Purity 98%
Chemical Name or Material Bathocuproin
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 225-240-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.