Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Batoclimab Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA542265
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542265 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542265 Supplier Invitrogen™ Supplier No. MA542265
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Batoclimab (HBM9161) is a fully human antibody targeting the neonatal Fc receptor (FcRn). It targets patients with neuromyelitis optica spectrum diseases and other autoimmune diseases, and can accelerate the degradation of autoantibodies in their bodies. Batoclimab (HBM9161) is an antibody that targets the neonatal Fc receptor (FcRn). FcRn expression is up-regulated by pro-inflammatory cytokines (such as TNF-alpha) and prolongs the half-life of IgG and serum albumin by reducing lysosomal degradation in endothelial cells and bone marrow-derived cells. Blocking the FcRn-IgG interaction can accelerate the degradation of autoantibodies and reduce the onset of various pathogenic IgG-mediated autoimmune diseases, including myasthenia gravis, thyroid ophthalmopathy, neuromyelitis optica spectrum disease and immunity Thrombocytopenia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Batoclimab
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 1.29 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias HL161
Host Species Human
Immunogen Human FCGRT.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 λ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.