Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bax Antibody (BAX/962), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP247815MFV450
Description
Bax Monoclonal antibody specifically detects Bax in Human, Monkey, Mouse (Negative), Rat (Negative) samples. It is validated for Flow CytometrySpecifications
Bax | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Monkey, Mouse (Negative), Rat (Negative) | |
Purified |
Flow Cytometry | |
BAX/962 | |
Flow Cytometry | |
apoptosis regulator BAX, BCL2-associated X protein, Bcl2-L-4, BCL2L4bcl2-L-4, Bcl-2-like protein 4 | |
Recombinant full-length human BAX protein. (Uniprot: Q07812) | |
0.1 mL | |
Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
581 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction