Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bax Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB10056095
Description
Bax Polyclonal specifically detects Bax in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Bax | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| apoptosis regulator BAX, BCL2-associated X protein, Bcl2-L-4, BCL2L4bcl2-L-4, Bcl-2-like protein 4 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Bax | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.05% Sodium Azide | |
| BAX | |
| A synthetic peptide corresponding to amino acids 43-61 (PELALDPVPQDASTKKLSE) of human Bax alpha was used as immunogen, GenBank no. NP_620116.1. The immunogen sequence is 100% conserved in human Bax isoforms alpha (192 amino acids), beta (218 amino acids), epsilon (164 amino acids), sigma (179 amino acids), and psi (173 amino acids). | |
| 0.05 mL | |
| Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
| 581 | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction